Please login to your online account to display your discounted pricing

BD 1063 dihydrochloride, Tocris

Selective σ1 ligand, putative antagonist

$164.61 - $678.85

Chemical Identifiers

Chemical Name or Material BD 1063 dihydrochloride
Assay Percent Range >99%
Molecular Formula C13H18Cl2N2·2HCl
Formula Weight 346.13
Synonym 1-[2-(3, 4-Dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

088310

 
r & d systems
088310
10mg Each for $164.61

088350

 
r & d systems
088350
50mg Each for $678.85
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 346.13
Chemical Name or Material BD 1063 dihydrochloride
Assay Percent Range >99%
Molecular Formula C13H18Cl2N2·2HCl
CAS 206996-13-6
Synonym 1-[2-(3, 4-Dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature