Please login to your online account to display your discounted pricing

BD 1047 dihydrobromide, Tocris


σ1 selective antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS095610

 View more versions of this product

Catalog No. 095610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 437.04
Chemical Name or Material BD 1047 dihydrobromide
Assay Percent Range >99%
CAS 138356-20-4
Molecular Formula C13H20Cl2N2·2HBr
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine dihydrobromide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material BD 1047 dihydrobromide
Assay Percent Range >99%
Molecular Formula C13H20Cl2N2·2HBr
Formula Weight 437.04
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine dihydrobromide
Accessories

Desiccate at +4°C