Please login to your online account to display your discounted pricing

BD 1047 dihydrobromide, Tocris

σ1 selective antagonist

$166.60 - $693.49

Chemical Identifiers

Chemical Name or Material BD 1047 dihydrobromide
Assay Percent Range >99%
Molecular Formula C13H20Cl2N2·2HBr
Formula Weight 437.04
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

095610

 
r & d systems
RDS095610
10mg Each for $166.60

095650

 
r & d systems
RDS095650
50mg Each for $693.49
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 437.04
Chemical Name or Material BD 1047 dihydrobromide
Assay Percent Range >99%
CAS 138356-20-4
Molecular Formula C13H20Cl2N2·2HBr
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine dihydrobromide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C