Please login to your online account to display your discounted pricing

BD 1008 dihydrobromide, Tocris


Potent, selective σ ligand

Quantity 10mg

Manufacturer: r & d systems  051110

 View more versions of this product

Catalog No. 051110

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material BD 1008 dihydrobromide
Quantity 10mg
CAS 138356-09-9
Recommended Storage Store at -20°C
Assay Percent Range >98%
Molecular Formula C15H22Cl2N2·2HBr
Formula Weight 463.08
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide

Specifications

Chemical Name or Material BD 1008 dihydrobromide
Assay Percent Range >98%
Molecular Formula C15H22Cl2N2·2HBr
Formula Weight 463.08
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide
Accessories

Store at -20°C