Please login to your online account to display your discounted pricing

BD 1008 dihydrobromide, Tocris

Potent, selective σ ligand

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material BD 1008 dihydrobromide
Assay Percent Range >98%
Molecular Formula C15H22Cl2N2·2HBr
Formula Weight 463.08
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

051110

 
r & d systems
051110
10mg Each for $181.19

051150

 
r & d systems
051150
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 463.08
CAS 138356-09-9
Chemical Name or Material BD 1008 dihydrobromide
Assay Percent Range >98%
Molecular Formula C15H22Cl2N2·2HBr
Synonym N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamine dihydrobromide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C