Please login to your online account to display your discounted pricing

BCTC, Tocris™

TRPV1 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material BCTC
Assay Percent Range >98%
Molecular Formula C20H25ClN4O
Formula Weight 372.89
Synonym 4-(3-Chloro-2-pyridinyl)-N-[4-(1, 1-dimethylethyl)phenyl]-1-piperazinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

387550

 
r & d systems
387550
50mg Each for $844.73

387510

 
r & d systems
387510
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 372.89
Quantity 50mg
Chemical Name or Material BCTC
Synonym 4-(3-Chloro-2-pyridinyl)-N-[4-(1, 1-dimethylethyl)phenyl]-1-piperazinecarboxamide
Recommended Storage Store at Room Temperature
CAS 393514-24-4
Assay Percent Range >98%
Molecular Formula C20H25ClN4O
Safety and Handling

Store at Room Temperature