missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bcl-XL Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

$282.27 - $1,320.53

Specifications

Antigen Bcl-XL Ab-2
Clone 7D9
Applications Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1334P Thermo Scientific™ Lab Vision™
MS1334P
Purified Ab with BSA and Azide 1mL Each for $1,205.41
MS1334P0 Thermo Scientific™ Lab Vision™
MS1334P0
Purified Ab with BSA and Azide 100μL Each for $282.27
MS1334P1 Thermo Scientific
MS1334P1
Purified Ab with BSA and Azide 500μL Each for $794.92
MS1334P1ABX Thermo Scientific™ Lab Vision™
MS1334P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $878.73
MS1334PABX Thermo Scientific™ Lab Vision™
MS1334PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,320.53
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-XL Ab-2, Mouse Monoclonal Antibody. 

 

Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family. Bcl-XL represses cell death, while Bcl-XS favors cell death.

Host Species: Mouse

Clone: 7D9

Isotype: IgG2a

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 35-50

Immunogen: A synthetic peptide corresponding to aa 35-50 (TEAPEETEAERETPSA) of mouse Bcl-XL protein

Molecular Weight: 27kDa

Positive Control: Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell/nuclear membrane.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Bcl-XL Ab-2
Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin),Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin)
Unlabeled
Murine
Apoptosis
A synthetic peptide corresponding to aa 35-50 (TEAPEETEAERETPSA) of mouse Bcl-xL protein
7D9
Monoclonal
Concentrated
RUO
Primary
IgG2a