Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bcl-XL Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS1334P1ABX

 View more versions of this product

Catalog No. MS1334P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Isotype IgG2a
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 35-50 (TEAPEETEAERETPSA) of mouse Bcl-xL protein
Antigen Bcl-XL Ab-2
Quantity 100µL
Format Concentrated
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Clone 7D9
Conjugate Unlabeled
Type Primary
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Apoptosis
Concentration 1mg/mL

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-XL Ab-2, Mouse Monoclonal Antibody. 

 

Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family. Bcl-XL represses cell death, while Bcl-XS favors cell death.

Host Species: Mouse

Clone: 7D9

Isotype: IgG2a

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 35-50

Immunogen: A synthetic peptide corresponding to aa 35-50 (TEAPEETEAERETPSA) of mouse Bcl-XL protein

Molecular Weight: 27kDa

Positive Control: Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell/nuclear membrane.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO