Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bcl-X Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-X Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS715P1ABX

 View more versions of this product

Catalog No. MS715P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Host Species Mouse
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Immunogen A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine bcl-X protein.
Clone 2H12
Format Concentrated
Isotype IgG2a
Quantity 100µL
Regulatory Status RUO
Antigen Bcl-X Ab-1
Conjugate Unlabeled
Research Discipline Apoptosis
Classification Monoclonal
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Type Primary

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-X Ab-1, Mouse Monoclonal Antibody.

Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family.

Host Species: Mouse

Clone: 2H12

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Pig. Others not tested.

Epitope: aa 3-14

Immunogen: A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine Bcl-X protein.

Molecular Weight: 27kDa

Positive Control: Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO