Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bcl-2α Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

$270.96 - $1,285.88

Specifications

Antigen Bcl-2alpha Ab-3
Clone 8C8
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS597B

 
thermo scientific™ lab vision™
MS597B
Biotinylated Ab with BSA and Azide 1mL Each for $1,285.88

MS597B0

 
thermo scientific™ lab vision™
MS597B0
Biotinylated Ab with BSA and Azide 100µL Each for $366.59

MS597B1

 
thermo scientific™ lab vision™
MS597B1
Biotinylated Ab with BSA and Azide 500µL Each for $872.55

MS597P

 
thermo scientific™ lab vision™
MS597P
Purified Ab with BSA and Azide 1mL Each for $1,009.57

MS597P0

 
thermo scientific™ lab vision™
MS597P0
Purified Ab with BSA and Azide 100µL Each for $270.96

MS597P1

 
thermo scientific™ lab vision™
MS597P1
Purified Ab with BSA and Azide 500µL Each for $663.90

MS597P1ABX

 
thermo scientific™ lab vision™
MS597P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $750.20

MS597PABX

 
thermo scientific™ lab vision™
MS597PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,080.90

MS597R7

 
thermo scientific
MS597R7
Ready-to-use for IHC (UVLP) 7mL Each for $347.37
Description & Specifications

Specifications

Antigen Bcl-2alpha Ab-3
Clone 8C8
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Host Species Mouse
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A Synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human bcl-2 protein
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments with Thermo Scientific™ Bcl-2α Ab-3, Mouse Monoclonal Antibody.

 

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse

Clone: 8C8

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with mouse and rat. Others not tested.

Epitope: aa 41-54

Immunogen: A synthetic peptide, aa 41-54 (Cys-GAAPAPGIFSSQPG) of human Bcl-2 protein

Molecular Weight: 25-26kDa

Positive Control: Raji or HeLa Cells, Tonsil or follicular lymphomas

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

<