Please login to your online account to display your discounted pricing

BBOT, Scintillation Grade, Acros Organics

$302.98 - $302.98

Chemical Identifiers

Chemical Name or Material BBOT
Assay ≥99.0%
Molecular Formula C26H26N2O2S
Formula Weight 430.57
MDL Number MFCD00005774
Synonym 2, 5-Bis(5'-tert-butyl-2-benzoxazol-2-yl)thiophene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC401731000

  SDS
thermo fisher scientific
401731000
Glass bottle 100g Each for $302.98
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 430.57
Physical Form Powder
Quantity 100g
Color Light yellow to green
RTECS Number DM4888332
EINECS Number 230-426-4
TSCA TSCA
Melting Point 199° to 202°C
Merck Index 15, 1012
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00005774
CAS 7128-64-5
Molecular Formula C26H26N2O2S
Chemical Name or Material BBOT
Suitability passes
Specific Extinction 47000 min. (372 -374nm)
Synonym 2, 5-Bis(5'-tert-butyl-2-benzoxazol-2-yl)thiophene

For fluorescence/luminescence spectroscopy