Please login to your online account to display your discounted pricing

(±)-Bay K 8644, Tocris

Ca2+-channel activator (L-type)

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material (±)-Bay K 8644
Assay Percent Range >99%
Molecular Formula C16H15F3N2O4
Formula Weight 356.3
Synonym 1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylic acid, methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

154410

 
r & d systems
RDS154410
10mg Each for $214.12

154450

 
r & d systems
RDS154450
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 356.3
Quantity 10mg
Chemical Name or Material (±)-Bay K 8644
Assay Percent Range >99%
Recommended Storage Store at +4°C
CAS 71145-03-4
Synonym 1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylic acid, methyl ester
Molecular Formula C16H15F3N2O4
Safety and Handling

Store at +4°C