Please login to your online account to display your discounted pricing

Bax Mouse, Clone: 6A7, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $259.55

Specifications

Antigen Bax
Clone 6A7
Host Species Murine
Species Reactivity Human, Monkey, Mouse, Rat
Applications Western Blot
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013580

 
life technologies
LS14699781
50μg Each for $158.05

5013581

 
life technologies
LS14699782
100μg Each for $259.55
Description & Specifications

Specifications

Antigen Bax
Clone 6A7
Host Species Murine
Species Reactivity Human, Monkey, Mouse, Rat
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 6A7 antibody reacts with mouse, rat, monkey and human Bax (20 kDa). The 6A7 antibody shows no cross-reaction with Bcl-2 or Bcl-xL proteins. Bcl-2, Bax, and Bcl-xL are members of a protein family that regulates apoptosis; while several family members including Bcl-2 and Bcl-xL inhibit apoptosis, overexpression of Bax promotes programmed cell death. A human Bax protein peptide sequence, aa 12-24, was used as immunogen.