Please login to your online account to display your discounted pricing

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

$268.92 - $1,494.51

Specifications

Antigen Bax Ab-4
Clone 6A7
Applications Immunofluorescence
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS714B

 
thermo scientific™
MS714B
200μg/mL Biotin 1mL Each for $1,494.51

MS714B0

 
thermo scientific™
MS714B0
200μg/mL Biotin 100μL Each for $363.96

MS714B1

 
thermo scientific™
MS714B1
200μg/mL Biotin 500μL Each for $1,000.94

MS714P

 
thermo scientific™
MS714P
200μg/mL Unlabeled 1mL Each for $1,160.08

MS714P0

 
thermo scientific™
MS714P0
200μg/mL Unlabeled 100μL Each for $268.92

MS714P1

 
thermo scientific™
MS714P1
200μg/mL Unlabeled 500μL Each for $760.96

MS714P1ABX

 
thermo scientific™
MS714P1ABX
1mg/mL Unlabeled 100μL Each for $845.59

MS714PABX

 
thermo scientific™
MS714PABX
1mg/mL Unlabeled 200μL Each for $1,258.02
Description & Specifications

Specifications

Antigen Bax Ab-4
Clone 6A7
Applications Immunofluorescence
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.
Isotype IgG1

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results. 

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 6A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: aa 12-24

Immunogen: A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: PC12 or HeLa cells

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO