Please login to your online account to display your discounted pricing

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody


Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™  MS714P1ABX

 View more versions of this product

Catalog No. MS714P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Isotype IgG1
Regulatory Status RUO
Clone 6A7
Quantity 100µL
Antigen Bax Ab-4
Immunogen A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.
Applications Immunofluorescence
Host Species Mouse
Conjugate Unlabeled
Research Discipline Apoptosis
Concentration 1mg/mL
Type Primary
Classification Monoclonal

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results. 

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 6A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: aa 12-24

Immunogen: A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: PC12 or HeLa cells

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO