Learn More
Bax Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.
Supplier: Thermo Fisher Scientific MS711PABX
Description
Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).Host Species: Mouse
Clone: 2D2
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: aa 3-16
Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.
Molecular Weight: 21kDa
Positive Control: HeLa cells, Hodgkin's lymphoma
Cellular Localization: Cytoplasmic and cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry (Not tested)
Note:
USA: RUO; Int'l: RUO
Specifications
Bax Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Apoptosis | |
Human | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2D2 | |
Unconjugated | |
A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein. | |
RUO | |
Primary | |
Concentrated |