Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Bax Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry,  western blotting, immunofluorescence, and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS711P1ABX

 View more versions of this product

Catalog No. MS711P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Bax Ab-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone 2D2
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein.
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments with Thermo Scientific™ Bax Ab-1, Mouse Monoclonal Antibody.

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 2D2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 3-16

Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: HeLa cells, Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Not tested)

Note:

USA: RUO; Int'l: RUO