missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bausch & Lomb™ Watchmaker Loupe Magnifiers

Available in a complete range of powers

Manufacturer:  Bausch & Lomb™ BL814171

 View more versions of this product

Catalog No. 12-032-14



Description

Description

  • Lightweight
  • Can be worn on headband if desired
  • Shell color: Black

Specifications

Specifications

Black
2.54cm
For magnification of fine, delicate work
3.81cm
Glass
1 in.
28 diopter
1.5 in.
7x
Watchmaker Loupe Magnifier
SDS
Documents