missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bausch & Lomb™ Watchmaker Loupe Magnifiers

Available in a complete range of powers

$33.34 - $42.50

Specifications

Color Black
Product Type Watchmaker Loupe Magnifier
For Use With (Application) For magnification of fine, delicate work
Material Glass
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (English) Outer Magnification Diopter Length (English) Focal Price Quantity  
Catalog Number Mfr. No. Diameter (English) Outer Magnification Diopter Length (English) Focal Price Quantity  
12-032-15
View Documents
Bausch & Lomb™
814173
1 in. 4x 16 diopter 2.5 in.
Each for $34.50
 
12-032-14
View Documents
Bausch & Lomb™
BL814171
1 in. 7x 28 diopter 1.5 in.
Each for $33.34
 
12-032-13
View Documents
Bausch & Lomb™
BL814170
1 in. 10x 40 diopter 1 in.
Each for $42.50
 
12-037E
View Documents
Bausch & Lomb™
BL814172
1 in. 5x 20 diopter 2 in.
Each for $35.03
 
Description

Description

  • Lightweight
  • Can be worn on headband if desired
  • Shell color: Black

Specifications

Specifications

Black
Watchmaker Loupe Magnifier
For magnification of fine, delicate work
Glass
SDS