missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bausch & Lomb™ Watchmaker Loupe Magnifiers

Available in a complete range of powers

Manufacturer:  Bausch & Lomb™ 814173

 View more versions of this product

Catalog No. 12-032-15



Description

Description

  • Lightweight
  • Can be worn on headband if desired
  • Shell color: Black

Specifications

Specifications

Black
2.54cm
For magnification of fine, delicate work
6.35cm
Glass
1 in.
16 diopter
2.5 in.
4x
Watchmaker Loupe Magnifier
SDS
Documents