missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bausch & Lomb™ Watchmaker Loupe Magnifiers

Available in a complete range of powers

Manufacturer:  Bausch & Lomb™ BL814170

 View more versions of this product

Catalog No. 12-032-13



Description

Description

  • Lightweight
  • Can be worn on headband if desired
  • Shell color: Black

Specifications

Specifications

Black
2.54cm
For magnification of fine, delicate work
2.54cm
Glass
1 in.
40 diopter
1 in.
10x
Watchmaker Loupe Magnifier
SDS
Documents
Product Certifications
Promotions

Promotions