Please login to your online account to display your discounted pricing

Bausch & Lomb™ Scales for Magnifiers

Dual ERK1/RasGAP inhibitor; Maintains ESC self-renewal

$104.88 - $104.88

Chemical Identifiers

Chemical Name or Material Pluripotin
CAS 839707-37-8
Assay Percent Range >98%
Molecular Formula C27H25F3N8O2
Formula Weight 550.54
Synonym N-[3-[7-[(1,3-Dimethyl-1H-pyrazol-5-yl)amino]-1,4-dihydro-1-methyl-2-oxopyrimido[4,5-d]pyrimidin-3(2H)-yl]-4-methylphenyl]-3-(trifluoromethyl)benzamide
View More Specs
Includes:  Protective storage pouch
Products
Catalog Number Mfr. No. Price Quantity    

BL813438

 
bausch & lomb inc
BL813438
Each for $104.88
Description & Specifications

Specifications

For Use With (Application) To measure 20mm in 0.1mm intervals
For Use With (Equipment) Lenscope™ magnifier
Includes Protective storage pouch
Material Glass
Product Type Metric Scale

  • Available in inch and metric versions
  • Inch scale measures 0.75 in. in 0.005 in. intervals
  • Metric scale measures 20mm in 0.1mm intervals