Please login to your online account to display your discounted pricing
Bausch & Lomb Lenscope™ Magnifiers
Ergonomic plastic body for easy handling
$179.47 - $179.47
Products
Catalog Number Mfr. No. Price Quantity    
12-038A
  Accessories
bausch & lomb inc
BL813442
Each for $179.47
Description & Specifications
  • Precision inspection tool with quality lens where a clear, distortion-free image is necessary
  • Two angled illumination lamps at 45° or 90° from the viewing angle for viewing in low-light conditions
  • Use for any close inspection with included lens or measurement application when you add and combine the measurement scales (sold separately)
  • Foam protection in case has precut secure areas to hold the measurement scale lenses as you add them to your set
  • Can be used with optional 12-037D scale
  • Interchangeable triplet lens sold as items 12-037D, 17-985-558 and BL-813438