missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bausch & Lomb™ Hastings Triplet Magnifiers

Three separate lenses bonded together to form a compound lens

Manufacturer:  Bausch & Lomb™ BL816175

 View more versions of this product

Catalog No. 12-032C



Includes: Protective pouch for storage

Description

Description

Three separate lenses bonded together to form a compound lens

  • Power viewing area presents a clean, sharp, crisp image with incorporated Bausch & Lomb precision optics
  • Provides images without distortion
  • Swing-away nickel-plated case protects magnifying glass, serves as handle and has key ring attachment
  • Magnification power and lens type engraved on nickel case for easy retrieval from storage or chain

Specifications

Specifications

0.51 in.
56 diopter
Protective pouch for storage
2.03cm
Glass
1.3cm
Used by professionals
0.8 in.
14X
Hastings Triplet Magnifier
SDS
Documents