Bausch & Lomb™ Hastings Triplet Adjustable Focus Measuring Magnifying Glass
Provides a flat image without distortion
$185.09 - $185.09
Specifications
Product Type Hastings Triplet Measuring Magnifier
For Use With (Application) To provide a flat image without distortion
Includes Protective case and storage pouch for case and lens
View More Specs
Includes: Protective case and storage pouch for case and lens
Products 1
Catalog Number Mfr. No. Magnification Diopter Length (English) Focal Length (Metric) Focal Price Quantity  
Catalog Number Mfr. No. Magnification Diopter Length (English) Focal Length (Metric) Focal Price Quantity  
12-037C
View Documents
Bausch & Lomb™
BL813435
7x 28 diopter 1.5 in. 3.9cm
Each for $185.09
Description
Description
  • Power viewing area presents a clean, sharp, crisp image with incorporated Bausch & Lomb precision optics
  • Lens adjusts for individual focus
  • Transparent body allows natural light illumination on the magnified area
  • Protective case keeps the magnifier safe, scratch free in all work environments
  • Scale sold separately (12-037D)
Specifications
Specifications
Hastings Triplet Measuring Magnifier
To provide a flat image without distortion
Protective case and storage pouch for case and lens
SDS