Bausch & Lomb™ Hastings Triplet Adjustable Focus Measuring Magnifying Glass
Provides a flat image without distortion
Manufacturer:  Bausch & Lomb™ BL813435
 View more versions of this product
Catalog No. 12-037C
Includes: Protective case and storage pouch for case and lens
Description
Description
  • Power viewing area presents a clean, sharp, crisp image with incorporated Bausch & Lomb precision optics
  • Lens adjusts for individual focus
  • Transparent body allows natural light illumination on the magnified area
  • Protective case keeps the magnifier safe, scratch free in all work environments
  • Scale sold separately (12-037D)
Specifications
Specifications
28 diopter
Protective case and storage pouch for case and lens
3.9cm
Glass
To provide a flat image without distortion
1.5 in.
7x
Hastings Triplet Measuring Magnifier