missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Batyl Alcohol, TCI America™

$169.59 - $1,480.45

Chemical Identifiers

CAS 544-62-7
Molecular Formula C21H44O3
Molecular Weight (g/mol) 344.58
MDL Number MFCD00004720
InChI Key OGBUMNBNEWYMNJ-UHFFFAOYSA-N
Synonym batyl alcohol, batilol, 1-o-octadecylglycerol, stearyl monoglyceride, batilolum, 3-octadecyloxy propane-1,2-diol, 3-octadecyloxy-1,2-propanediol, glycerine 1-monostearyl ether, monooctadecyl ether of glycerol, batylalcohol
PubChem CID 3681
ChEBI CHEBI:34117
IUPAC Name 3-octadecoxypropane-1,2-diol
SMILES CCCCCCCCCCCCCCCCCCOCC(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B120225G View Documents TCI America
B120225G
25g Each for $169.59
 
B1202500G View Documents TCI America
B1202500G
500g Each for $1,480.45
 
Specifications

Chemical Identifiers

544-62-7
344.58
OGBUMNBNEWYMNJ-UHFFFAOYSA-N
3681
3-octadecoxypropane-1,2-diol
C21H44O3
MFCD00004720
batyl alcohol, batilol, 1-o-octadecylglycerol, stearyl monoglyceride, batilolum, 3-octadecyloxy propane-1,2-diol, 3-octadecyloxy-1,2-propanediol, glycerine 1-monostearyl ether, monooctadecyl ether of glycerol, batylalcohol
CHEBI:34117
CCCCCCCCCCCCCCCCCCOCC(CO)O

Specifications

White
25g
C21H44O3
batyl alcohol, batilol, 1-o-octadecylglycerol, stearyl monoglyceride, batilolum, 3-octadecyloxy propane-1,2-diol, 3-octadecyloxy-1,2-propanediol, glycerine 1-monostearyl ether, monooctadecyl ether of glycerol, batylalcohol
CCCCCCCCCCCCCCCCCCOCC(CO)O
344.58
CHEBI:34117
Crystal-Powder at 20°C
Batyl Alcohol
71°C
544-62-7
MFCD00004720
OGBUMNBNEWYMNJ-UHFFFAOYSA-N
3-octadecoxypropane-1,2-diol
3681
344.58
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0414

RTECSNumber : TZ0960000

TSCA : Yes