missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Batimastat, Tocris

Potent, broad spectrum MMP inhibitor

$168.15 - $362.13

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C23H31N3O4S2
Formula Weight 477.64
Synonym (2R, 3S)-N4-Hydroxy-N1-[(1S)-2-(methylamino)-2-oxo-1-(phenylmethyl)ethyl]-2-(2-methylpropyl)-3-[(2-thienylthio)methyl]butanediamide
Chemical Name or Material Batimastat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
29-611 R & D Systems
29611
1mg Each for $168.15
29-611-0 R & D Systems
296110
10mg Each for $362.13
Specifications Description & Specifications

Chemical Identifiers

>99%
477.64
Batimastat
C23H31N3O4S2
(2R, 3S)-N4-Hydroxy-N1-[(1S)-2-(methylamino)-2-oxo-1-(phenylmethyl)ethyl]-2-(2-methylpropyl)-3-[(2-thienylthio)methyl]butanediamide

Specifications

130370-60-4
C23H31N3O4S2
(2R, 3S)-N4-Hydroxy-N1-[(1S)-2-(methylamino)-2-oxo-1-(phenylmethyl)ethyl]-2-(2-methylpropyl)-3-[(2-thienylthio)methyl]butanediamide
Batimastat
>99%
477.64
1mg

Safety and Handling

Recommended Storage : Store at -20°C