missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC164050050

 View more versions of this product

Catalog No. AC164050050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Bathophenanthrolinedisulfonic acid, disodium salt hydrate
Glass bottle
4% max. (K.F.)
>300°C
52746-49-3
100.0
2NaO3S·xH2O
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+]
536.5
536.5
98%
Authentic
suitable for determination of iron
Beige-Pink
5g
97.5
98%
MFCD00149310
PFNPMHXHQCVXED-UHFFFAOYSA-L
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
54669730
Powder

Chemical Identifiers

52746-49-3
536.5
PFNPMHXHQCVXED-UHFFFAOYSA-L
54669730
C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+]
2NaO3S·xH2O
MFCD00149310
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : 258-152-

SDS Documents