Learn More
Bathocuproin sulfonate disodium salt hydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B22550MD
Description
Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.
Solubility
Miscible with water.
Notes
Incompatible with sulfuric acid, sodium metal, potassium metal and calcium carbide.
Specifications
Bathocuproin sulfonate disodium salt hydrate | |
>300°C | |
MFCD00149974 | |
9033637 | |
Miscible with water. | |
[Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O | |
564.54 | |
564.55 (Anhydrous) |
52698-84-7 | |
C26H18N2Na2O6S2 | |
250 mg | |
disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
RNGKZLRAVYPLJC-UHFFFAOYSA-L | |
disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
15678335 | |
97% |
Chemical Identifiers
52698-84-7 | |
564.54 | |
RNGKZLRAVYPLJC-UHFFFAOYSA-L | |
15678335 |
C26H18N2Na2O6S2 | |
MFCD00149974 | |
disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate | |
[Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 258-111-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only