Learn More
Bathocuproin, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2284106
Description
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.
Solubility
Soluble in methanol (36mg/100ml), ethanol, 2-propanol, bezene, and acetone. Partly miscible in water.
Notes
Store in cool, dry conditions in a sealed container. Store away from oxidizing agents.
Specifications
Bathocuproin | |
279°C to 285°C | |
MFCD00004972 | |
306714 | |
Soluble in methanol (36mg/100ml),ethanol,2-propanol,bezene,and acetone. Partly miscible in water. | |
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1 | |
360.46 | |
360.46 |
4733-39-5 | |
C26H20N2 | |
5 g | |
bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563 | |
STTGYIUESPWXOW-UHFFFAOYSA-N | |
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline | |
65149 | |
98% |
Chemical Identifiers
4733-39-5 | |
360.46 | |
STTGYIUESPWXOW-UHFFFAOYSA-N | |
65149 | |
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1 |
C26H20N2 | |
MFCD00004972 | |
bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563 | |
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 225-240-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only