missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BATF Mouse anti-Human, PE, Clone: MBM7C7, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $421.08

Specifications

Antigen BATF
Clone MBM7C7
Host Species Murine
Gene Alias B cell activating transcription factor, ATF-like
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245542 Life Technologies
LS12986041
25 Tests Each for $170.28
50245543 Life Technologies
LS12986042
100 Tests Each for $421.08
Specifications

Description

The MBM7C7 monoclonal antibody reacts with human and mouse BATF (B cell activating transcription factor). BATF is a 14kDa member of the AP-1 family of transcription factors and is involved in the repression of other AP-1 family members. BATF is upregulated in activated T cells when compared to naïve T and B cells and has been shown to be critical for Th17, but not Th1 or Th2 differentiation. In Th17, BATF interacts with the promoters of IL17, IL17F, IL21 and IL22. BATF has also been shown to be involved in CD8 T cell exhaustion in HIV infection.
Description & Specifications

Specifications

BATF
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MBM7C7
B cell activating transcription factor, ATF-like
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test