Please login to your online account to display your discounted pricing

Basophil Marker Mouse anti-Human, eFluor 450, Clone: 2D7, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $462.55

Specifications

Antigen Basophil Marker
Clone 2D7
Host Species Murine
Gene Alias 2D7
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124227

 
life technologies
LS48974841
25 Tests Each for $201.55

501124228

 
life technologies
LS48974842
100 Tests Each for $462.55
Description & Specifications

Specifications

Antigen Basophil Marker
Clone 2D7
Host Species Murine
Gene Alias 2D7
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 2D7 monoclonal antibody recognizes human 2D7 antigen expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining. This antibody does not stain mast cells.