missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SP Scientific Basket and Cover

Designed for use in SP Scientific Hotpack Glass Washers; Undercounter, Freestanding and Stacking models. Glass Washer Accessories maximize space, improve efficiency and protect specialty glassware.

Manufacturer:  SP Scientific 437127

Catalog No. 15365224



Description

Description

  • 7.5 L x 7.5 W x 4 in.D
  • Stainless Steel
Specifications

Specifications

open basket and lid
Stainless Steel
7.5 in.
1
4 in.
10cm
Accessory
0.45kg
19cm
7.5 in. x 7.5 in.
Washing Lab Glassware
Stainless Steel
19cm
Lab Glassware Washer
19cm
7.5 in.
1#
7.5 in.
7.5 in. x 7.5 in. x 4 in.
Basket
SDS
Documents