Please login to your online account to display your discounted pricing

Fisher Scientific™ Basix™ Filter Tips


The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Manufacturer: fisher scientific™  A23013SERV

 View more versions of this product

Catalog No. 13-611-104

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Length (Metric) 3.17cm
Length (English) 1.25 in.
For Use With (Equipment) Research grade pipetters
Product Type Basix™ Universal Fit Filter Tip
Sterility Sterile
Packaging Rack
Color Natural
Certifications/Compliance RNase, DNase, DNA, Pyrogen free
Volume (Metric) 10μL
Autoclavable No
Tip Style Filter

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.

Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.