Please login to your online account to display your discounted pricing

Fisher Scientific™ Basix™ Filter Tips


The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Manufacturer: fisher scientific™  F109RSSERV

 View more versions of this product

Catalog No. 13-611-122

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Basix™ Universal Fit Filter Tip
Volume (Metric) 300μL
Sterility Sterile
Color Natural
Autoclavable No
Tip Style Filter
Packaging Rack
Length (English) 2.312 in.
Length (Metric) 5.87cm
Certifications/Compliance RNase, DNase, DNA, Pyrogen free
For Use With (Equipment) Research grade pipetters

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.

Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.