Please login to your online account to display your discounted pricing

Fisher Scientific™ Basix™ Filter Tips


The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Manufacturer: fisher scientific™  F110NXLRSERV

 View more versions of this product

Catalog No. 13-611-114

  • / Pack of 1632 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Basix™ Universal Fit Filter Tip
Volume (Metric) 200μL
Sterility Sterile
Color Natural
Autoclavable No
Tip Style Filter
Packaging Rack
Length (English) 3.52 in.
Length (Metric) 8.94cm
Certifications/Compliance RNase, DNase, DNA, Pyrogen free
For Use With (Equipment) Research grade pipetters

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.

Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.