Please login to your online account to display your discounted pricing

Basic pH Meter


Ideal for classroom use

Manufacturer: fisher scientific  840088

 View more versions of this product

Catalog No. S96285

  • / Each
 

Description & Specifications

Specifications

Includes Basic pH meter, Epoxy body sealed reference combination electrode with a BNC connector and a 2.5 ft. cable, pH buffers (4 and 7), deionized water, 9V battery and instructions housed in a slim protective hardshell, Foam lined case
Range (pH) 0 to 14
Resolution (pH) 0.01
Accuracy (pH) ±0.07
Calibration Points (pH) 2
Display Type LCD
Battery Type Alkaline
Certifications/Compliance CE
Weight (Metric) 0.23kg
Detection Range 0 to 14pH
For Use With (Application) pH
Voltage 9V