Please login to your online account to display your discounted pricing
Basic pH Meter
Ideal for classroom use
$402.30 - $402.30
Specifications
Accuracy (pH) ±0.07
Battery Type Alkaline
Calibration Points (pH) 2
Certifications/Compliance CE
Display Type LCD
View More Specs
Includes:  Meter supplied with 9V battery, calibration tool, tripod screw, and full instructions. Meter Kit includes basic pH meter, epoxy-body sealed reference combination electrode with a BNC connector and a 2.5 ft. cable, pH buffers (4 and 7), deionized water, 9V battery and instructions housed in a slim protective hardshell, foam-lined case.
Products
Catalog Number Mfr. No. Includes Weight (Metric) Price Quantity    
S96285
fisher scientific
840088
Basic pH meter, Epoxy body sealed reference combination electrode with a BNC connector and a 2.5 ft. cable, pH buffers (4 and 7), deionized water, 9V battery and instructions housed in a slim protective hardshell, Foam lined case 0.23kg Each for $402.30
Description & Specifications
Specifications
Accuracy (pH) ±0.07
Battery Type Alkaline
Calibration Points (pH) 2
Certifications/Compliance CE
Display Type LCD
Range (pH) 0 to 14
Resolution (pH) 0.01
For Use With (Application) pH
  • Large 7/8 in. high digits and lightweight design make this pH meter ideal for both field and lab applications
  • Simple to operate and works with any electrode with a BNC connector
  • Measures the full 0 to 14 pH range with a high 0.01 pH resolution
  • Features two-point calibration, hold function, fold out easel back and a tripod screw
  • Also available in a complete, ready-to-use kit for immediate field use
CE marked