Please login to your online account to display your discounted pricing

Basic Fuchsin, pure, certified, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  401690250

 View more versions of this product

Catalog No. AC401690250

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Dark green
Physical Form Glistening Crystalline Powder
Formula Weight 337.84
Quantity 25g
Health Hazard 3 Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Harmful if swallowed. May cause eye, skin, and respiratory tract irritation.
NFPA
Purity Grade Pure
Beilstein 13, 765
Merck Index 15, 5715
Synonym Basic Violet 14, hydrochloride
Chemical Name or Material Basic Fuchsin
Dye Content 88% min.
CAS 632-99-5
Name Note Pure
Molecular Formula C20H19N3·HCl
Health Hazard 1 Warning
MDL Number MFCD00012569
Packaging Glass bottle
Infrared Spectrum Authentic
Absorption Ratio 1.16 to 1.35
Lambda Max 548 to 552nm
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.

Specifications

Chemical Name or Material Basic Fuchsin
Name Note Pure
CAS 632-99-5
Purity Grade Pure
Molecular Formula C20H19N3·HCl
Formula Weight 337.84
MDL Number MFCD00012569
Synonym Basic Violet 14, hydrochloride
Accessories

Warning

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Harmful if swallowed. May cause eye, skin, and respiratory tract irritation.
NFPA

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.

SDS & Safety Documents