Please login to your online account to display your discounted pricing

Base Oil 75, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 1 gal.
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  BASE75G

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. BASE-75G

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1 gal.
Health Hazard 1 Causes skin irritation, serious eye irritation
Chemical Name or Material Base Oil 75 Blank
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Nonhazardous
Solvent or Matrix Base Oil 75
Certificate of Analysis With Certificate of Analysis

Specifications

Chemical Name or Material Base Oil 75 Blank
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation

Ambient