missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Base Oil 20, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc BASE20

 View more versions of this product

Catalog No. BASE-20


 

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As(+5)1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Description & Specifications

Specifications

500mL
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Base Oil 20 Blank
With Certificate of Analysis.
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Base Oil 20

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Base Oil 20 Blank

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient