Please login to your online account to display your discounted pricing

Base/Neutrals Surrogate Standard, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 5mL
Packaging 5mL amber prescored glass ampule

Manufacturer: spex certiprep llc  CLP90-SB5

 View more versions of this product

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. SPEX-N-022

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5mL
Health Hazard 1 Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.
Chemical Name or Material Base/Neutrals Surrogate Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
CAS 1718-51-0
CAS 2199-69-1
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 321-60-8
Packaging 5mL amber prescored glass ampule
Recommended Storage Ambient
Analyte or Component Names 1,2-Dichlorobenzene-d4,2-Fluorobiphenyl
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS 4165-60-0
DOT Information Nonhazardous
Solvent or Matrix MeCl:Acetone

Specifications

Chemical Name or Material Base/Neutrals Surrogate Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Dichlorobenzene-d42199-69-11000μg/mL
2-Fluorobiphenyl321-60-81000μg/mL
Nitrobenzene-d54165-60-01000μg/mL
p-Terphenyl-d141718-51-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.

Ambient