Thermo Scientific™ Barnstead™ RO Storage Reservoirs
Benefit from automatic operation with these polythylene and fiberglass storage reservoirs for Thermo Scientific™ Barnstead™ reverse osmosis water systems.
$2,601.11 - $2,601.11
Specifications
For Use With For RO water storage
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Price Quantity  
09-034-184 Thermo Scientific
D9021
16 gal. Each for $2,601.11
Specifications
Description
Benefit from automatic operation with polythylene and fiberglass Thermo Scientific™ Barnstead™ RO Storage Reservoirs. For use with Barnstead RO (Reverse Osmosis) water systems.
Polyethylene Reservoirs:
  • Control panel that displays reservoir's water level
  • Vent protected to eliminate the introduction of airborne contamination
  • Draw-off points that include spigot, dishwasher port (for 60 and 100L reservoirs only), and feed to Thermo Scientific™ Nanopure™ systems
  • Can be wall- or bench-mounted
  • Constructed of rigid fluorinated polyethylene, providing strength and protection against material contamination of the water
  • Pump protector provided; ensures the system is not damaged when there is not enough water in the reservoir
Single Access Port:
  • Single port to connect reservoir to other systems and/or equipment
  • Capacity: 30L
Multiple Access Port:
  • Optional distribution pump available
  • Perfect for supplying water to a remote Nanopure or glassware washer: easily mounts to base of 16 gal. (60L) reservoir and can supply reverse osmosis water up to 35′ (10.7m) away
  • Capable of delivering up to 1 gal. (3.8L) of water/min. at 15psig (1.3 bar)
  • Automatically circulates water for 15 minutes every hour when in standby
  • Multiple ports to connect reservoir to other systems and/or equipment
Recommended for
  • RO water storage
Description & Specifications
Specifications
For RO water storage