Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilters
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
Supplier: Thermo Scientific™ 50133981
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
- GenPure™†
- GenPure™ Pro†
- GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
- MicroPure™‡
- Smart2 Pure™‡
‡UF and UF/UV units only
Specifications
Ultrafilter; For use with MicroPure, Smart2 Pure (UF and UF/UV units only) | |
MicroPure, Smart2Pure (UF and UF/UV units only) | |
Ultrafilter |