Please login to your online account to display your discounted pricing

Thermo Scientific™ Barnstead™ Cabinetized Stills

Save space and installation time with a Thermo Scienitific™ Barnstead™ Cabinetized Still, a compact, all-in-one water distillation solution.

Specifications

Auto Drain At 4-hr. intervals
View More Specs
Products
Catalog Number Mfr. No. Amperage Price Quantity    

09-028-41A

 
thermo scientific™
A1065C
17A CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Auto Drain At 4-hr. intervals

Save space and installation time with a Thermo Scienitific™ Barnstead™ Cabinetized Still, an all-in-one water distillation solution. This solution includes a still and an appropriately matched tin-coated storage tank in a compact storage cabinet. Systems are fully automatic and designed for point-of-use installation. Produce water pure enough for biological and biochemical procedures that are sensitive to even minute traces of contamination.

Features:

  • Stills come in choice of two sizes: 2 or 5 gal/hr.
  • Q-Baffle ensures high-quality, pyrogen-free product water.
  • Fully automatic operation allows for unattended use.
  • Still turns off when tank is full and restarts when tank empties.
  • Still automatically drains boiling chamber every time the unit is off, and at four-hour intervals of operation.

Model 210 Still—2 gal./hr. with 10 gal. Tank:

  • Purity meter that alerts user of distilled water purity in storage tank
  • Includes low water cut-off for safety protection

Model 525 Still—5 gal./hr. with 25 gal. Tank:

  • Purity meter that alerts user of distilled water purity in storage tank
  • Optional recirculation pump and base mounting stand available
  • Storage tank is equipped with a UV lamp for improved bacterial control

Ordering Information:

Optional accessories include Bench Stand (A1066, for Model 210 only), Floor Stand (A1521, for Model 525 only), and Recirculation Pump with Floor Stand (A1522, for Model 525 only); all sold separately. Contact your sales representative for details.

This product is not available in all regions.

Recommended for:

Buffer/Reagent preparation, glassware washing, dishwasher feed