missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium zirconium oxide, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1402830

 View more versions of this product

Catalog No. AA1402830



Specifications

Specifications

Barium zirconium oxide
99% (Metal basis)
MFCD00014189
barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
[O-][Zr](=O)[O-].[Ba+2]
276.548
276.53
5.52g/cm3
White
12009-21-1
BaO3Zr
Off-white powder
DQBAOWPVHRWLJC-UHFFFAOYSA-N
barium(2+);dioxido(oxo)zirconium
16212444
Powder
Odorless
250g

Chemical Identifiers

12009-21-1
276.548
DQBAOWPVHRWLJC-UHFFFAOYSA-N
16212444
[O-][Zr](=O)[O-].[Ba+2]
BaO3Zr
MFCD00014189
barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
barium(2+);dioxido(oxo)zirconium
Safety and Handling

Safety and Handling

H302-H332

EINECSNumber : 234-546-8

Documents