Learn More
Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014028A1
Description
Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium zirconium oxide | |
12009-21-1 | |
Powder | |
99% (Metal basis) | |
MFCD00014189 | |
Insoluble in water. | |
[O-][Zr](=O)[O-].[Ba+2] | |
276.548 | |
276.53 | |
Off-white powder |
2500°C | |
White | |
1 kg | |
BaO3Zr | |
barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis | |
DQBAOWPVHRWLJC-UHFFFAOYSA-N | |
barium(2+);dioxido(oxo)zirconium | |
16212444 | |
Odorless | |
5.52 |
Chemical Identifiers
12009-21-1 | |
276.548 | |
DQBAOWPVHRWLJC-UHFFFAOYSA-N | |
16212444 | |
[O-][Zr](=O)[O-].[Ba+2] |
BaO3Zr | |
MFCD00014189 | |
barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis | |
barium(2+);dioxido(oxo)zirconium |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 234-546-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only