Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium zirconium isopropoxide, 10% w/v in isopropanol

Precursor of barium zirconium oxide using Sol-Gel method

Chemical Identifiers

Chemical Name or Material Barium zirconium isopropoxide
Formula Weight 583.08
MDL Number MFCD00798551
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41047AC

 
alfa aesar™
41047AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41047AE

 
alfa aesar™
41047AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 583.08
Physical Form Liquid
Chemical Name or Material Barium zirconium isopropoxide
MDL Number MFCD00798551
Sensitivity Moisture Sensitive
UN Number UN3274
Health Hazard 1 Danger
Concentration 10% w/v in isopropanol
Quantity 25mL
Health Hazard 2 GHS H Statement
H225-H302-H332-H336
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H302-H332-H336
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.