Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium yttrium tungsten oxide anhydrous, 99.9%

Chemical Identifiers

Chemical Name or Material Barium yttrium tungsten oxide
CAS 37265-86-4
Name Note anhydrous
Molecular Formula Ba3Y3WO9
Formula Weight 1006.58g/mol
MDL Number MFCD00213929
UN Number UN1564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1748318

 
alfa aesar™
A17483-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Chemical Name or Material Barium yttrium tungsten oxide
Name Note anhydrous
CAS 37265-86-4
Assay 99.9%
Molecular Formula Ba3Y3WO9
Formula Weight 1006.58g/mol
MDL Number MFCD00213929
UN Number UN1564
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.